5-(Hydroxymethyl)furan-2-carboxylic Acid ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
H1750-1G
H1750-5G
TCIAH1750-1GEA
35
EUR
TCIAH1750-1G
TCIAH1750-5G
5-(Hydroxymethyl)furan-2-carboxylic Acid ≥98.0% (by GC, titration analysis)
5-(Hydroxymethyl)furan-2-carboxylic Acid
Formula:
C6H6O4 Temperatura de almacenaje: Nevera |
Número MDL:
MFCD03274472 Núm. CAS: 6338-41-6 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 159.0 to 163.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...