3-Hydroxyisobenzofuran-1(3H)-one ≥98.0% (by GC, titration analysis)
Proveedor: TCI
Sinónimos:
3-Hydroxyphthalide, 3-Hydroxy-2-benzofuran-1(3H)-one, 3-Hydroxy-1(3H)-isobenzofuranone
H1754-25G
H1754-5G
TCIAH1754-25GEA
157
EUR
TCIAH1754-25G
TCIAH1754-5G
3-Hydroxyisobenzofuran-1(3H)-one ≥98.0% (by GC, titration analysis)
3-Hydroxyisobenzofuran-1(3H)-one
Formula:
C₈H₆O₃ MW: 150,13 g/mol |
Número MDL:
MFCD00956142 Núm. CAS: 16859-59-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 98.0 to 102.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...