Inosine 5'-monophosphate disodium salt hydrate ≥98.0% (by HPLC and UV method)

Proveedor: TCI

Sinónimos: 5'-IMP 2Na hydrate, 5'-Inosinic acid disodium salt hydrate, Inosinic acid (disodium)(hydrate)(1:2:X), Inosine 5'-monophosphate disodium salt xhydrate, Disodium inosinate hydrate, Inosic acid disodium salt hydrate

I0036-1G I0036-25G I0036-5G
TCIAI0036-1GEA 16 EUR
TCIAI0036-1G TCIAI0036-25G TCIAI0036-5G
Inosine 5'-monophosphate disodium salt hydrate ≥98.0% (by HPLC and UV method)
Inosine 5'-monophosphate disodium salt hydrate
Formula: C₁₀H₁₁N₄Na₂O₈P·xH₂O
MW: 392,17 g/mol (anhy)
Número MDL: MFCD00150372
Núm. CAS: 352195-40-5
EINECS: 225-146-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3155c0497ae7766a4c054a039890e6f82561dfb9&vid=3155c0497ae7766a4c054a039890e6f82561dfb9&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (UV-vis method) min. 97.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water max. 29.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR