Isohexane (mixture of isomeric branched chain Hexanes) ≥98.0% (por GC)
Proveedor: TCI
|
Peligro
|
Sinónimos:
Hexanes
I0134-25ML
I0134-500ML
TCIAI0134-25MLEA
16
EUR
TCIAI0134-25ML
TCIAI0134-500ML
Isohexane (mixture of isomeric branched chain Hexanes) ≥98.0% (por GC)
Isohexane (mixture of isomeric branched chain Hexanes)
Formula:
C₆H₁₄ MW: 86.18 g/mol |
Número MDL:
MFCD02179311 Núm. CAS: 73513-42-5 UN: 1208 ADR: 3 II |
Resultados test especificaciones
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0% (total of isomers) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...