Isoproterenol hydrochloride ≥99.0% (by titrimetric analysis)

Proveedor: TCI

I0260-25G I0260-5G
TCIAI0260-25GEA 231 EUR
TCIAI0260-25G TCIAI0260-5G
Isoproterenol hydrochloride ≥99.0% (by titrimetric analysis)
Isoproterenol hydrochloride
Formula: C₁₁H₁₇NO₃
Número MDL: MFCD00012603
Núm. CAS: 51-30-9
EINECS: 200-089-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 99.0 %
Solubility in Water very faint turbidity

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR