Iopanoic acid ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: 3-Amino-alpha-ethyl-2,4,6-triiodohydrocinnamic Acid, 3-(3-Amino-2,4,6-triiodophenyl)-2-ethylpropionic Acid, Iodopanoic Acid, 2-(3-Amino-2,4,6-triiodobenzyl)butyric Acid

I0300-25G I0300-5G
TCIAI0300-25GEA 751 EUR
TCIAI0300-25G TCIAI0300-5G
Iopanoic acid ≥98.0% (by titrimetric analysis)
Iopanoic acid
Formula: C₁₁H₁₂I₃NO₂
MW: 570,94 g/mol
Número MDL: MFCD00038687
Núm. CAS: 96-83-3
EINECS: 202-539-9
Índice Merck: 14,05058

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 150.0 to 156.0 deg-C
Solubility in 1mol/L NaOH within almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR