Sodium-2-naphthalenesulphonate ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
I0353-25G
I0353-5G
TCIAI0353-25GEA
58
EUR
TCIAI0353-25G
TCIAI0353-5G
Sodium-2-naphthalenesulphonate ≥98.0% (by HPLC, titration analysis)
Sodium-2-naphthalenesulphonate
Formula:
C₁₀H₈O₃S |
Número MDL:
MFCD00064186 Núm. CAS: 532-02-5 EINECS: 208-523-8 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Ion exchange titration) | min. 98.0 % (calcd.on anh.substance) |
Absorbance (380nm) | max. 0.250 |
Absorbance (390nm) | max. 0.150 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...