(R)-2-Isopropylamino-2-phenylethanol ≥98.0% (by GC, titration analysis)

Proveedor: TCI

Sinónimos: (R)-2-(Isopropylamino)-2-phenylethanol

I0476-25G I0476-5G I0476-1G
TCIAI0476-25GEA 637 EUR
TCIAI0476-25G TCIAI0476-5G TCIAI0476-1G
(R)-2-Isopropylamino-2-phenylethanol ≥98.0% (by GC, titration analysis)
(R)-2-Isopropylamino-2-phenylethanol
Formula: C₁₁H₁₇NO
MW: 179,26 g/mol
Número MDL: MFCD00671548
Núm. CAS: 112211-92-4

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 75.0 to 79.0 deg-C
Specific rotation [a]20/D -65.0 to -73.0 deg (C=1,ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR