Isopropyl methyl sulphone ≥97.0% (por GC)

Proveedor: TCI

I0484-25G I0484-5G
TCIAI0484-25GEA 259 EUR
TCIAI0484-25G TCIAI0484-5G
Isopropyl methyl sulphone ≥97.0% (por GC)
Isopropyl methyl sulphone
Formula: C₄H₁₀O₂S
MW: 122,18 g/mol
Número MDL: MFCD00050415
Núm. CAS: 4853-74-1

Order Now

Resultados test especificaciones

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR