2-Iodobenzenesulphonic acid, hidrato ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Peligro
|
I0758-1G
I0758-5G
TCIAI0758-1GEA
78
EUR
TCIAI0758-1G
TCIAI0758-5G
2-Iodobenzenesulphonic acid, hidrato ≥98.0% (by HPLC, titration analysis)
2-Iodobenzenesulphonic acid
Formula:
C₆H₅IO₃S MW: 284,07 g/mol (anhy) |
Número MDL:
MFCD01860232 Núm. CAS: 63059-25-6 UN: 3261 ADR: 8,III |
Resultados test especificaciones
Appearance | White to Almost white powder to crystaline |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % (calcd.on anh.substance) |
Water | 6.0 to 13.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...