2-(tert-Butylamino)-4-(cyclopropylamino)-6-(methylthio)-1,3,5-triazine ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Cybutryne, N2-(tert-Butyl)-N4-cyclopropyl-6-(methylthio)-1,3,5-triazine-2,4-diamine
I0842-5G
I0842-25G
TCIAI0842-5GEA
48
EUR
TCIAI0842-5G
TCIAI0842-25G
2-(tert-Butylamino)-4-(cyclopropylamino)-6-(methylthio)-1,3,5-triazine ≥98.0% (by titrimetric analysis)
2-(tert-Butylamino)-4-(cyclopropylamino)-6-(methylthio)-1,3,5-triazine
Formula:
C₁₁H₁₉N₅S MW: 253,37 g/mol |
Número MDL:
MFCD01863779 Núm. CAS: 28159-98-0 EINECS: 248-872-3 UN: 3077 ADR: 9,III |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 126.0 to 130.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...