3-((2'-(1H-Tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)-2-butyl-1,3-diazaspiro[4.4]non-1-en-4-one ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
Irbesartan, 3-((2'-(1H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-2-butyl-1,3-diazaspiro[4.4]non-1-en-4-one
I0859-1G
I0859-5G
TCIAI0859-1GEA
16
EUR
TCIAI0859-1G
TCIAI0859-5G
3-((2'-(1H-Tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)-2-butyl-1,3-diazaspiro[4.4]non-1-en-4-one ≥98.0% (by HPLC, titration analysis)
3-((2'-(1H-Tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)-2-butyl-1,3-diazaspiro[4.4]non-1-en-4-one
Formula:
C₂₅H₂₈N₆O MW: 428,5294 g/mol |
Número MDL:
MFCD00864464 Núm. CAS: 138402-11-6 Índice Merck: 14,05083 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 184.0 to 188.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...