Ácido 4-iodo-3-nitrobenzoico ≥97.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
I0980-1G
I0980-5G
TCIAI0980-1GEA
16
EUR
TCIAI0980-1G
TCIAI0980-5G
Ácido 4-iodo-3-nitrobenzoico ≥97.0% (by GC, titration analysis)
Ácido 4-iodo-3-nitrobenzoico
Formula:
C₇H₄INO₄ MW: 293.02 g/mol |
Número MDL:
MFCD00502399 Núm. CAS: 35674-27-2 |
|
Resultados test especificaciones
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 210.0 to 214.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...