Imidazole hydrobromide, low water content ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Imidazolium bromide
I1006-5G
I1006-1G
TCIAI1006-5GEA
167
EUR
TCIAI1006-5G
TCIAI1006-1G
Imidazole hydrobromide, low water content ≥98.0% (by HPLC, titration analysis)
Imidazole hydrobromide
Formula:
C₃H₄N₂·HBr MW: 148.99 g/mol Pto. de fusión: 221°C |
Núm. CAS:
101023-55-6 |
|
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Water | max. 100 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...