5-Iodo-o-toluidina ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
5-Iodo-2-metilanilina
I1021-5G
I1021-25G
TCIAI1021-5GEA
49
EUR
TCIAI1021-5G
TCIAI1021-25G
5-Iodo-o-toluidina ≥98.0% (by GC, titration analysis)
5-Iodo-o-toluidina
Formula:
C₇H₈IN MW: 233,05 g/mol |
Número MDL:
MFCD00051531 Núm. CAS: 83863-33-6 EINECS: 281-094-2 UN: 2811 ADR: 6.1,III |
Resultados test especificaciones
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 48.0 to 52.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...