Inulin

Proveedor: TCI

Sinónimos: Dahlin, Alantin, Inulin (by Enzymatic synthesis)

I1067-25G I1067-500G
TCIAI1067-25GEA 18 EUR
TCIAI1067-25G TCIAI1067-500G
Inulin
Inulin
Número MDL: MFCD00131407
Núm. CAS: 9005-80-5
EINECS: 232-684-3
Índice Merck: 14,05004

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Drying loss max. 10.0 %
Ignition residue (Sulfate) max. 0.5 %
Dextrin and Starch to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR