6H-Indolo[2,3-b]quinoxaline ≥98.0% (by GC, titration analysis)

Proveedor: TCI

I1098-1G I1098-200MG
TCIAI1098-1GEA 120 EUR
TCIAI1098-1G TCIAI1098-200MG
6H-Indolo[2,3-b]quinoxaline ≥98.0% (by GC, titration analysis)
6H-Indolo[2,3-b]quinoxaline
Formula: C₁₄H₉N₃
Número MDL: MFCD00068754
Núm. CAS: 243-59-4

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR