3,4-Dihydro-4-oxo-1,2,3-benzotriazine ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-Ketobenzotriazine, 1,2,3-Benzotriazine-4(3H)-one, Benzo[d][1,2,3]triazin-4(3H)-one
K0003-25G
K0003-5G
TCIAK0003-25GEA
57
EUR
TCIAK0003-25G
TCIAK0003-5G
3,4-Dihydro-4-oxo-1,2,3-benzotriazine ≥98.0% (by HPLC, titration analysis)
3,4-Dihydro-4-oxo-1,2,3-benzotriazine
Formula:
C₇H₅N₃O MW: 147,14 g/mol |
Número MDL:
MFCD00052387 Núm. CAS: 90-16-4 EINECS: 201-971-5 |
Resultados test especificaciones
Appearance | White to Light red to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in N,N-DMF | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...