Kinetin ≥99.0% (by HPLC, titration analysis)

Proveedor: TCI

K0009-1G K0009-5G
TCIAK0009-1GEA 23 EUR
TCIAK0009-1G TCIAK0009-5G
Kinetin ≥99.0% (by HPLC, titration analysis)
Kinetin
Formula: C₁₀H₉N₅O
Número MDL: MFCD00075757
Núm. CAS: 525-79-1
EINECS: 208-382-2
Índice Merck: 14,05311

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Nonaqueous Titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR