Calcium-3-methyl-2-oxovalerate hydrate ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: 3-Methyl-2-oxovaleric Acid Calcium Salt

K0022-25G K0022-5G
TCIAK0022-25GEA 304 EUR
TCIAK0022-25G TCIAK0022-5G
Calcium-3-methyl-2-oxovalerate hydrate ≥98.0% (by titrimetric analysis)
Calcium-3-methyl-2-oxovalerate hydrate
Formula: C₆H₁₀O₃
MW: 298.35 g/mol
Número MDL: MFCD00002583
Núm. CAS: 66872-75-1
EINECS: 266-505-5

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Water max. 11.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR