Calcium-4-methyl-2-oxovalerate hydrate ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
4-Methyl-2-oxovaleric Acid Calcium Salt
K0023-25G
K0023-5G
TCIAK0023-25GEA
295
EUR
TCIAK0023-25G
TCIAK0023-5G
Calcium-4-methyl-2-oxovalerate hydrate ≥98.0% (by titrimetric analysis)
Calcium-4-methyl-2-oxovalerate hydrate
Formula:
C₆H₁₀O₃ MW: 298,35 g/mol |
Número MDL:
MFCD00004163 Núm. CAS: 51828-95-6 EINECS: 257-457-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Chelometric Titration) | min. 98.0 % (calcd.on anh.substance) |
Water | max. 12.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...