(S)-(+)-Ketopinic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: (S)-(+)-Ketopinic Acid

K0028-1G K0028-5G
TCIAK0028-1GEA 61 EUR
TCIAK0028-1G TCIAK0028-5G
(S)-(+)-Ketopinic acid ≥98.0% (by GC, titration analysis)
(S)-(+)-Ketopinic acid
Formula: C₁₀H₁₄O₃
Número MDL: MFCD00168053
Núm. CAS: 40724-67-2

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 235.0 to 240.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR