Kojic acid dipalmitate ≥98.0% (by HPLC)
Proveedor: TCI
Sinónimos:
Pentadecyl 4-oxo-6-((palmitoyloxy)methyl)-4H-pyran-3-carboxylate, 5-(Palmitoyloxy)-2-[(palmitoyloxy)methyl]-4H-pyran-4-one
K0050-5G
K0050-25G
TCIAK0050-5GEA
22
EUR
TCIAK0050-5G
TCIAK0050-25G
Kojic acid dipalmitate ≥98.0% (by HPLC)
Kojic acid dipalmitate
Formula:
C₃₈H₆₆O₆ MW: 618,93 g/mol |
Número MDL:
MFCD03840587 Núm. CAS: 79725-98-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 94.0 to 98.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...