Ketorolac tromethamine ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

Sinónimos: Ketorolac tris salt, 2-Amino-2-(hydroxymethyl)propane-1,3-diol 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylate, Ketorolac tromethamine salt

K0053-1G K0053-5G
TCIAK0053-1GEA 45 EUR
TCIAK0053-1G TCIAK0053-5G
Ketorolac tromethamine ≥98.0% (by HPLC, titration analysis)
Ketorolac tromethamine
Formula: C₁₅H₁₃NO₃·C₄H₁₁NO₃
MW: 376,4037 g/mol
Número MDL: MFCD00887595
Núm. CAS: 74103-07-4
UN: 2811
ADR: 6.1,III
Índice Merck: 14,05306
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65c7e850db2b2a4015bfbcd31b50392f67bc30bc&vid=65c7e850db2b2a4015bfbcd31b50392f67bc30bc&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR