Lauric anhydride ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
L0017-25G
L0017-500G
L0017-100G
TCIAL0017-25GEA
45
EUR
TCIAL0017-25G
TCIAL0017-500G
TCIAL0017-100G
Lauric anhydride ≥98.0% (by titrimetric analysis)
Lauric anhydride
Formula:
C₂₄H₄₆O₃ |
Número MDL:
MFCD00008965 Núm. CAS: 645-66-9 EINECS: 211-449-9 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Morpholine Method) | min. 98.0 % |
Melting point | 40.0 to 43.0 deg-C |
Solubility in Acetone | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...