DL-Leucyl-DL-phenylalanine ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: H-DL-Leu-DL-Phe-OH

L0038-1G L0038-100MG
TCIAL0038-1GEA 84 EUR
TCIAL0038-1G TCIAL0038-100MG
DL-Leucyl-DL-phenylalanine ≥98.0% (by titrimetric analysis)
DL-Leucyl-DL-phenylalanine
Formula: C₁₅H₂₂N₂O₃
MW: 278,35 g/mol
Número MDL: MFCD00037258
Núm. CAS: 56217-82-4

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in 1mol/L HCl almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR