Sodium ligninsulphonate

Proveedor: TCI

Sinónimos: Ligninsulphonic acid Sodium Salt

L0098-500G L0098-25G
TCIAL0098-500GEA 60 EUR
TCIAL0098-500G TCIAL0098-25G
Sodium ligninsulphonate
Sodium ligninsulphonate
Número MDL: MFCD00135572
Núm. CAS: 8061-51-6

Order Now

Resultados test especificaciones

Appearance Orange to Brown powder to crystal
Methoxyl Group 5.0 to 7.0 % (calcd.on anh.substance)
Water max. 6.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR