D-Lysine methyl ester dihydrochloride ≥98.0% (by total nitrogen and titration analysis)

Proveedor: TCI

Sinónimos: H-D-Lys-OMe.2HCl, (R)-Methyl 2,6-diaminohexanoate dihydrochloride

L0201-25G L0201-5G
TCIAL0201-25GEA 157 EUR
TCIAL0201-25G TCIAL0201-5G
D-Lysine methyl ester dihydrochloride ≥98.0% (by total nitrogen and titration analysis)
D-Lysine methyl ester dihydrochloride
Formula: C₇H₁₆N₂O₂
MW: 233,13 g/mol
Pto. de fusión: 205°C
Número MDL: MFCD00039067
Núm. CAS: 67396-08-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe57501e2c5966c2420c933d40e281d910e41e0d&vid=527d88a3c07c7fdc655d2707c2238f808108033c&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D -14.0 to -19.0 deg (C=5, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR