Lac-β(1-4)lac-β-C10 ≥98.0% (by HPLC)

Proveedor: TCI

L0229-10MG L0229-100MG
TCIAL0229-10MGEA 89 EUR
TCIAL0229-10MG TCIAL0229-100MG
Lac-β(1-4)lac-β-C10 ≥98.0% (by HPLC)
Lac-beta(1-4)lac-beta-C10
Formula: C₃₄H₆₂O₂₁
Temperatura de almacenaje: Congelador
Núm. CAS: 1858224-00-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR