Levetiracetam ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Atención
|
Sinónimos:
(S)-2-(2-Oxopyrrolidin-1-yl)butanamide, (S)-2-(2-Oxo-1-pyrrolidinyl)butyramide
L0234-1G
L0234-5G
TCIAL0234-1GEA
52
EUR
TCIAL0234-1G
TCIAL0234-5G
Levetiracetam ≥98.0% (by HPLC, total nitrogen)
Levetiracetam
Formula:
C₈H₁₄N₂O₂ MW: 170,21 g/mol |
Número MDL:
MFCD03265610 Núm. CAS: 102767-28-2 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 116.0 to 120.0 deg-C |
Specific rotation [a]20/D | -86.0 to -90.0 deg (C=1, Acetone) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...