Lithium trifluoro(trifluoromethyl)borate - dimethyl carbonate complex ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
L0240-1G
L0240-5G
TCIAL0240-1GEA
152
EUR
TCIAL0240-1G
TCIAL0240-5G
Lithium trifluoro(trifluoromethyl)borate - dimethyl carbonate complex ≥98.0% (by titrimetric analysis)
Lithium trifluoro(trifluoromethyl)borate - dimethyl carbonate complex
Formula:
CBF₆Li Temperatura de almacenaje: Congelador |
Núm. CAS:
1443685-69-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...