Lomerizine dihydrochloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

Sinónimos: 1-[Bis(4-fluorophenyl)methyl]-4-(2,3,4-trimethoxybenzyl)piperazine Dihydrochloride

L0245-200MG L0245-1G
TCIAL0245-200MGEA 45 EUR
TCIAL0245-200MG TCIAL0245-1G
Lomerizine dihydrochloride ≥98.0% (by HPLC, titration analysis)
Lomerizine dihydrochloride
Formula: C₂₇H₃₀F₂N₂O₃
MW: 541,46 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD01703867
Núm. CAS: 101477-54-7
Índice Merck: 14,05563
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceb95c327a28875a134641c652d2c0000288a2de&vid=ceb95c327a28875a134641c652d2c0000288a2de&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR