Lithium bis(fluorosulphonyl)imide ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

Sinónimos: LiFSI, Lithium bis(fluorosulphonyl)amide

L0281-25G L0281-5G
TCIAL0281-25GEA 259 EUR
TCIAL0281-25G TCIAL0281-5G
Lithium bis(fluorosulphonyl)imide ≥98.0% (by titrimetric analysis)
Lithium bis(fluorosulphonyl)imide
Formula: F₂LiNO₄S₂
MW: 187,06 g/mol
Número MDL: MFCD28126786
Núm. CAS: 171611-11-3
UN: 1759
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=544971c92ffdb96d7b113816b99fb65bb73e21bd&vid=544971c92ffdb96d7b113816b99fb65bb73e21bd&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR