Levomefolic acid ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: (S)-2-(4-((((S)-2-Amino-5-methyl-4-oxo-3,4,5,6,7,8-hexahydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, (S)-2-[4-[[[(S)-2-Amino-5-methyl-4-oxo-1,4,5,6,7,8-hexahydropteridin-6-yl]methyl]amino]benzamido]pentanedioic acid, N-[4-[[((6S)-2-Amino-5-methyl-4-oxo-1,4,5,6,7,8-hexahydro-6-pteridinyl)methyl]amino]benzoyl]-L-glutamic acid

L0335-250MG L0335-50MG
TCIAL0335-250MGEA 357 EUR
TCIAL0335-250MG TCIAL0335-50MG
Levomefolic acid ≥98.0% (by HPLC)
Levomefolic acid
Formula: C₂₀H₂₅N₇O₆
Número MDL: MFCD00870135
Núm. CAS: 31690-09-2

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR