DL-alpha-Lipoic acid-NHS ≥96.0% (by HPLC)
Proveedor: TCI
Sinónimos:
Thioctic acid N-Hydroxysuccinimide ester, 5-(1,2-Dithiolan-3-yl)pentanic acid 2,5-Dioxopyrrolidin-1-yl ester, α-Lipoic acid-NHS, 2,5-Dioxopyrrolidin-1-yl 5-(1,2-Dithiolan-3-yl)pentanoate
L0345-50MG
L0345-250MG
TCIAL0345-50MGEA
111
EUR
TCIAL0345-50MG
TCIAL0345-250MG
DL-alpha-Lipoic acid-NHS ≥96.0% (by HPLC)
DL-alpha-Lipoic acid-NHS
Formula:
C₁₂H₁₇NO₄S₂ Temperatura de almacenaje: Congelador |
Número MDL:
MFCD09028032 Núm. CAS: 40846-94-4 |
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) (CAD) | min. 96.0 area% |
Melting point | 93.0 to 97.0 deg-C |
Elemental analysis (Nitrogen) | 5.40 to 5.90 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...