Sodium-2-mercaptobenzothiazole ≥97.0% (by HPLC)

Proveedor: TCI
Atención

M0057-25G M0057-500G
TCIAM0057-25GEA 16 EUR
TCIAM0057-25G TCIAM0057-500G
Sodium-2-mercaptobenzothiazole ≥97.0% (by HPLC)
Sodium-2-mercaptobenzothiazole
Formula: C₇H₅NS₂
Número MDL: MFCD00065334
Núm. CAS: 2492-26-4
EINECS: 219-660-8
UN: 3077
ADR: 9,III

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to granule
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 93.0 %
Drying loss max. 6.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR