Methyl mercaptan sodium salt ∼15% in water

Proveedor: TCI
Peligro

Sinónimos: Sodium methanethiolate, Sodium thiomethoxide, Sodium methylsulphanide, Methanethiol sodium salt

M0096-25G M0096-500G
TCIAM0096-25GEA 19 EUR
TCIAM0096-25G TCIAM0096-500G
Methyl mercaptan sodium salt ∼15% in water
Methyl mercaptan sodium salt
Formula: CH₃NaS
MW: 70.08 g/mol
Pto. de inflamación: 50°C
Número MDL: MFCD00174316
Núm. CAS: 5188-07-8
EINECS: 225-969-9
UN: 2920
ADR: 8 II
Índice Merck: 14,05956

Order Now

Resultados test especificaciones

Appearance Colorless to Light orange to Yellow clear liquid
Concentration (Iodometric titration) 14.5 to 16.5 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR