Methylandrostenediol ≥98.0% (por GC)

Proveedor: TCI

M0146-1G M0146-25G
TCIAM0146-1GEA 31 EUR
TCIAM0146-1G TCIAM0146-25G
Methylandrostenediol ≥98.0% (por GC)
Methylandrostenediol
Formula: C₂₀H₃₂O₂
Número MDL: MFCD00021126
Núm. CAS: 521-10-8
EINECS: 208-301-0
Índice Merck: 14,05950

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 202.0 to 207.0 deg-C
Specific rotation [a]20/D -71.0 to -77.0 deg (C=1, EtOH)
Solubility in EtOH very faint turbidity

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR