(Methyleneamino)acetonitrile

Proveedor: TCI
Atención

Sinónimos: alpha-Hydroformamine Cyanide, Methyleneglycinonitrile

M0212-25G M0212-500G
TCIAM0212-25GEA 23 EUR
TCIAM0212-25G TCIAM0212-500G
(Methyleneamino)acetonitrile
(Methyleneamino)acetonitrile
Formula: C₃H₄N₂
MW: 68.08 g/mol
Número MDL: MFCD00001889
Núm. CAS: 109-82-0
EINECS: 203-709-5
Índice Merck: 14,06057

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 128.0 to 131.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR