Methyl hesperidine ≥90.0% (by UV/Vis)

Proveedor: TCI

M0338-25G M0338-5G
TCIAM0338-25GEA 91 EUR
TCIAM0338-25G TCIAM0338-5G
Methyl hesperidine ≥90.0% (by UV/Vis)
Methyl hesperidine
Formula: C₂₉H₃₆O₁₅
Número MDL: MFCD01741310
Núm. CAS: 11013-97-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddc663f559f480e18eab828159d0b704c9df8db0&vid=ddc663f559f480e18eab828159d0b704c9df8db0&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (UV-vis method) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR