N-Methylsaccharin ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
2-Methylbenzo[d]isothiazol-3(2H)-one 1,1-dioxide
M0427-25G
M0427-5G
TCIAM0427-25GEA
230
EUR
TCIAM0427-25G
TCIAM0427-5G
N-Methylsaccharin ≥98.0% (por GC)
N-Methylsaccharin
Formula:
C₈H₇NO₃S MW: 197,21 g/mol |
Número MDL:
MFCD00059263 Núm. CAS: 15448-99-4 EINECS: 239-466-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 130.0 to 134.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...