Sodium myristate ≥98.0% (by GC, titration analysis)

Proveedor: TCI

M0483-25G M0483-500G M0483-100G
TCIAM0483-25GEA 31 EUR
TCIAM0483-25G TCIAM0483-500G TCIAM0483-100G
Sodium myristate ≥98.0% (by GC, titration analysis)
Sodium myristate
Formula: C₁₄H₂₈O₂
Número MDL: MFCD00042653
Núm. CAS: 822-12-8
EINECS: 212-487-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d0b3634823875db4ae575c44b324eeefc10648c&vid=1d0b3634823875db4ae575c44b324eeefc10648c&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in hot EtOH (50vol%) almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR