Variamine blue B ≥98.0% (by titrimetric analysis) indicador redox
Proveedor: TCI
|
Atención
|
Sinónimos:
4-Amino-4'-methoxydiphenylamine Hydrochloride, N-(4-Methoxyphenyl)-1,4-phenylenediamine Hydrochloride, Variamine Blue B Hydrochloride
M0494-1G
M0494-25G
TCIAM0494-1GEA
16
EUR
TCIAM0494-1G
TCIAM0494-25G
Variamine blue B ≥98.0% (by titrimetric analysis) indicador redox
Variamine blue B
Formula:
C₁₃H₁₄N₂O MW: 250,73 g/mol |
Número MDL:
MFCD00012989 Núm. CAS: 3566-44-7 EINECS: 222-652-7 |
Resultados test especificaciones
Appearance | Blue to Dark blue powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Solubility in Water | very faint turbidity |
Drying loss | max. 0.5 % |
Sensitiveness | Colorimetric Test : Turn Blue purple with 1ppm Iron and turn Colorless to Almost colorless by adding EDTA 2Na |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...