DL-Methionine methylsulphonium chloride ≥99.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: Vitamin U, (3-Amino-3-carboxypropyl)dimethylsulphonium chloride

M0644-25G M0644-500G
TCIAM0644-25GEA 26 EUR
TCIAM0644-25G TCIAM0644-500G
DL-Methionine methylsulphonium chloride ≥99.0% (by titrimetric analysis)
DL-Methionine methylsulphonium chloride
Formula: C₆H₁₄ClNO₂S
MW: 199,69 g/mol
Número MDL: MFCD00031700
Núm. CAS: 3493-12-7
EINECS: 222-484-4
Índice Merck: 14,06098

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 99.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR