Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers) ≥80.0% (por GC)

Proveedor: TCI
Peligro

Sinónimos: Methylnadic Anhydride, Methylnadic anhydride, Nadic Methyl Anhydride, Methyl-3,6-endomethylenetetrahydrophthalic Anhydride

M0690-25G M0690-500G
TCIAM0690-25GEA 16 EUR
TCIAM0690-25G TCIAM0690-500G
Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers) ≥80.0% (por GC)
Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers)
Formula: C₁₀H₁₀O₃
Pto. de inflamación: 135°C
Número MDL: MFCD00167036
Núm. CAS: 25134-21-8
EINECS: 246-644-8
UN: 3265
ADR: 8,III

Order Now

Resultados test especificaciones

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 80.0 % (total of isomer)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR