4,4'-Methylenebis(cyclohexylamine) (mixture of isomers) ≥97.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
4,4'-Diaminodicyclohexylmethane, 4,4-Diaminodicyclohexyl methane, 4,4'-Methylenebis(aminocyclohexane), Bis(4-aminocyclohexyl)methane
M0699-25G
M0699-500G
TCIAM0699-25GEA
18
EUR
TCIAM0699-25G
TCIAM0699-500G
4,4'-Methylenebis(cyclohexylamine) (mixture of isomers) ≥97.0% (by GC, titration analysis)
4,4'-Methylenebis(cyclohexylamine) (mixture of isomers)
Formula:
C₁₃H₂₆N₂ MW: 210,37 g/mol Pto. Ebullición: 126°C |
Número MDL:
MFCD00001496 Núm. CAS: 1761-71-3 EINECS: 217-168-8 UN: 2928 ADR: 6.1,II |
Resultados test especificaciones
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 97.0 % (total of isomer) |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...