2-Methoxy-2,4-diphenyl-3(2H)-furanone ≥98.0% (by HPLC)

Proveedor: TCI

M0722-1G M0722-100MG
TCIAM0722-1GEA 423 EUR
TCIAM0722-1G TCIAM0722-100MG
2-Methoxy-2,4-diphenyl-3(2H)-furanone ≥98.0% (by HPLC)
2-Methoxy-2,4-diphenyl-3(2H)-furanone
Formula: C₁₇H₁₄O₃
MW: 266,30 g/mol
Número MDL: MFCD00036834
Núm. CAS: 50632-57-0

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 94.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR