DL-O-Methylserine ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: 2-Amino-3-methoxypropanoic acid

M0888-1G M0888-5G
TCIAM0888-1GEA 100 EUR
TCIAM0888-1G TCIAM0888-5G
DL-O-Methylserine ≥98.0% (by titrimetric analysis)
DL-O-Methylserine
Formula: C₄H₉NO₃
MW: 119,12 g/mol
Número MDL: MFCD00037773
Núm. CAS: 19794-53-7

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR