4-Methoxy-4'-nitrobiphenyl ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
4-(4'-Nitrophenyl)anisole, 4-(4'-Methoxyphenyl)nitrobenzene, 4-Methoxy-4'-nitro-1,1'-biphenyl
M0997-1G
M0997-100MG
TCIAM0997-1GEA
203
EUR
TCIAM0997-1G
TCIAM0997-100MG
4-Methoxy-4'-nitrobiphenyl ≥98.0% (por GC)
4-Methoxy-4'-nitrobiphenyl
Formula:
C₁₃H₁₁NO₃ MW: 229,24 g/mol |
Número MDL:
MFCD00143334 Núm. CAS: 2143-90-0 |
Resultados test especificaciones
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 108.0 to 111.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...