trans-1-Methyl-4-carboxy-5-(3-pyridyl)-2-pyrrolidinone ≥95.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
M1053-1G
M1053-100MG
M1053-5G
TCIAM1053-1GEA
49
EUR
TCIAM1053-1G
TCIAM1053-100MG
TCIAM1053-5G
trans-1-Methyl-4-carboxy-5-(3-pyridyl)-2-pyrrolidinone ≥95.0% (by GC, titration analysis)
trans-1-Methyl-4-carboxy-5-(3-pyridyl)-2-pyrrolidinone
Formula:
C₁₁H₁₂N₂O₃ |
Número MDL:
MFCD00075394 Núm. CAS: 161171-06-8 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 192.0 to 196.0 deg-C |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...